Learn More
Waters Corp Waters SunFire Prep Guard Cartridge, C18, 10 μm, 100 Å, 30 x 10 mm
Supplier:  Waters Corp 186006884
Encompass_Preferred
This preparative guard cartridge is designed to protect both HPLC and SFC 30 mm ID preparative columns. Placing this guard assembly ahead of the preparative column helps extend column lifetimes and reduce purification costs.
  • C18 chemistry
  • 10 μm particle size
  • 100 Å pore diameter
  • 30 mm inner diameter
  • 10 mm length
  • Reversed-Phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Designed to protect HPLC and SFC columns
Catalog No. 50-203-7742