Learn More
Waters Corp SunFire Sentry Guard Cartridge, C18, 3.5 μm, 4.6 mm, 20 mm
Supplier: Waters Corp 186002682
This Sentry Guard Cartridge is designed for general purpose method development at low pH. The C18 ligand provides high retention, particularly for basic compounds, making it ideal for purification and impurity profile assays.
- Chemistry: C18
- Separation mode: Reversed phase
- Particle substrate: Silica
- pH range: 2 to 8
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped
- Low silanol activity
- Spherical particle shape
- Particle size: 3.5 μm
- Pore size: 100 Å
- Designed for HPLC systems
- USP classification: L1
- Inner diameter: 4.6 mm
- Length: 20 mm
- Carbon load: 16%
- Two units per package
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.