Learn More
Waters Corp Waters SunFire C8 Column, 3.5 μm, 100 Å, 2.1 mm x 150 mm
Supplier:  Waters Corp 186002712
Encompass_Preferred
SunFire C8 columns are designed for general purpose method development at low pH. The C8 ligand is less hydrophobic and thus less retentive compared to SunFire C18, making it suitable for certain assays.
  • Used for general purpose method development at low pH.
  • C8 ligand is less hydrophobic and less retentive compared to a C18 column, preferred for some assays.
Catalog No. 50-781-303