Learn More
Waters Corp Waters SunFire C8 Column, 100Å, 3.5 μm, 2.1 mm x 100 mm
Supplier: Waters Corp 186002711
SunFire C8 columns are utilized for general purpose method development in low pH conditions. The C8 ligand is favored in certain assays due to its lower hydrophobicity, resulting in less retention when compared to SunFire C18.
- Chemistry: C8
- Separation mode: Reversed phase
- Particle substrate: Silica
- pH range: 2 to 8
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped: Yes
- Silanol activity: Low
- Particle shape: Spherical
- Particle size: 3.5 μm
- Pore size: 100 Å
- Format: Column
- Surface area: 340
- System: HPLC
- USP classification: L7
- Inner diameter: 2.1 mm
- Length: 100 mm
- Carbon load: 12 %
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.