Learn More
Waters Corp Waters Column, C8, 100Å, 3.5 μm, 2.1 mm x 50 mm
Supplier: Waters Corp 186002710
SunFire C8 columns are designed for general purpose method development in low pH conditions. The C8 ligand offers less hydrophobicity and, consequently, less retention compared to SunFire C18 columns, making it a preferred choice for specific assays.
- C8 chemistry for specific assays.
- Reversed phase separation mode.
- Silica particle substrate.
- PH range of 2 to 8.
- Maximum pressure of 6000 psi (415 Bar).
- Endcapped.
- Low silanol activity.
- Spherical particle shape.
- Particle size of 3.5 μm.
- Endfitting type.
- Pore size of 100 Å.
- Column format.
- Surface area of 340.
- HPLC system compatible.
- USP classification L7.
- Inner diameter of 2.1 mm.
- Length of 50 mm.
- Carbon load of 12 %.
- 1 unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.