Learn More
Waters Corp Waters Column, C8, 100Å, 5μm, 2.1 mm X 50 mm
Supplier:  Waters Corp 186002715
Encompass_Preferred
These C8 columns are designed for general purpose method development at low pH. Being less hydrophobic than C18 columns, they are preferred for some assays. They offer sharp peaks, remarkable sensitivity, high peak capacity, and ultra-low bleed for various mass spectrometry applications, providing great resolution, minimal backpressure, and speed, saving time and expense during analysis.
  • Longer lifetime due to low pH stability.
  • Patented synthesis, bonding, end-capping, and packing ensure high efficiency and sensitivity.
  • Provides consistent chromatographic results throughout its lifetime, maintaining exceptional repeatability.
  • Made with high purity raw materials under strict quality control.
  • Recommended with a VanGuard Cartridge to remove contaminants; made with the same materials as the analytical column.
Catalog No. 50-781-306