Learn More
Waters Corp HPLC Column, C8, 5 μm, 4.6 mm ID, 30 mm L
Supplier:  Waters Corp 186002734
Encompass_Preferred
SunFire C8 columns are utilized for general-purpose method development under low pH conditions. The C8 ligand is favored for certain assays due to its reduced hydrophobicity, making it less retentive when compared to SunFire C18 columns.
  • Endcapped
  • Spherical
  • Low silanization activity
Catalog No. 50-781-325