Learn More
Waters Corp Column, SunFire C8 OBD, 5 μm, 10 x 100 mm
Supplier:  Waters Corp 186008159
Encompass_Preferred
Waters SunFire C8 OBD Preparative Columns are engineered for efficient and robust lab-scale separations. Featuring a patented Optimum Bed Density (OBD) design, these columns ensure uniform packing and consistent performance, enabling direct scalability from analytical to preparative chromatography with high recoveries and extended column lifetimes.
  • Excellent peak shape and minimal column bleed for clean baselines.
  • High mass loading capacity for greater sample volume.
  • Reproducible performance with narrow specification ranges.
  • Wide pH operating range (2–8) for C8 chemistry.
  • Resistant to mechanical chromatographic bed failure, ensuring stability.
  • Enables fast, efficient separations and direct scalability from analytical methods.
Catalog No. 50-239-1588