Learn More
Waters Corp Waters Prep Guard Cartridge, C18, 10 μm, 100 Å
Supplier: Waters Corp 186002663
SunFire C18 columns are designed for general-purpose method development at low pH. The C18 ligand provides high retention, particularly for basic compounds. It is well-suited for purification and impurity profile assays.
- Chemistry: C18
- Separation mode: Reversed Phase
- Particle substrate: Silica
- pH range: 2 to 8 pH
- Temperature limits: 45°C
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped: Yes
- Silanol activity: Low
- Particle shape: Spherical
- Particle size: 10 μm
- Pore size: 100 Å
- Format: Prep Guard Cartridge
- Surface area: 340
- System: HPLC
- Technique: LC/MS, LC
- USP classification: L1
- Inner diameter: 10 mm
- Length: 10 mm
- Carbon load: 16%
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.