Learn More
Waters Corp Waters Prep Guard Cartridge, C18, 100Å, 10 μm, 19 mm X 10 mm
Supplier:  Waters Corp 186002666
Encompass_Preferred
SunFire C18 columns are designed for general purpose method development at low pH. They provide high retention for basic compounds. This Prep Guard Cartridge extends column longevity by preventing suspended particles and contaminants. It’s ideal for biological fluid cleanup.
  • Low pH method development
  • High retention for basic compounds
  • Purification and impurity assays
  • Extends column longevity
  • Prevents suspended particles and contaminants
  • High-performance packing materials
  • Ideal for biological fluid cleanup
  • Manufactured in cGMP, ISO 9001 facility
  • Batch tested chromatographically
Catalog No. 50-781-270