Learn More
Waters Corp Waters OBD Prep Column, C18, 5 μm, 100 Å, 50 mm L x 50 mm ID
Supplier:  Waters Corp 186002867
Encompass_Preferred
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
  • Uniform, optimally packed column beds for ideal outcomes in preparative chromatography.
  • Direct scalability from analytical to preparative LC.
  • Excellent column lifetime.
  • Consistent column-to-column performance.
  • Good resolution, peak shape, and mass loading.
  • On-target results for successful Prep LC purification with OBD Technology.
  • General purpose, balanced non-polar retention for acids, bases, and neutrals.
  • Excellent loading.
Catalog No. 50-781-410
Add to Cart
Request a Quote