Learn More
Waters Corp HPLC Guard Cartridge, SunFire C8, 100Å 5 μm, 10 × 10 mm
Supplier: Waters Corp 186002750
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
- Used for general purpose method development at low pH.
- C8 ligand is less hydrophobic and less retentive compared to C18, suitable for specific assays.
- Carbon load: 12.0%.
- Endcapping: Yes.
- Inner diameter: 10 mm.
- Mode: Reversed Phase.
- Particle size: 5.0 μm.
- pH range: 2 to 8.
- Pore size: 100 Å.
- USP number: L7.
- Length: 10 mm.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.