Learn More
Waters Corp Waters Corporation SunFire C8 OBD Prep Column, 50 x 150 mm, 10 μm
Supplier:  Waters Corp 186002767
Encompass_Preferred
The SunFire C8 OBD Prep Column is designed for preparative liquid chromatography, offering uniform and optimally packed column beds. This ensures excellent column lifetime and consistent performance, facilitating direct scalability from analytical to preparative applications. Its C8 chemistry provides less retentivity than a C18 column, making it suitable for strongly hydrophobic compounds.
  • Helps bridge the performance gap in Prep LC with uniform, optimum packed bed density.
  • Direct scalability from analytical to preparative chromatography.
  • Excellent column lifetime and consistent column-to-column performance.
  • Delivers good resolution, peak shape, and mass loading for preparative chromatography results.
  • Shorter C8 alkyl chain provides less retentivity than a typical C18, suitable for strongly hydrophobic compounds.
Catalog No. 50-781-358
Add to Cart
Request a Quote