Learn More
Waters Corp Waters SunFire C8 OBD Prep Column, 100Å, 10 μm, 30 x 50 mm
Supplier:  Waters Corp 186003853
Encompass_Preferred
The SunFire C8 OBD Prep Column offers efficient, scalable, lab-scale purifications with high recovery and extended column lifetimes, leveraging robust particles and the patented Optimum Bed Density (OBD) design.
  • Increased productivity with higher recoveries and longer column lifetimes.
  • Fast, efficient, lab-scale separations for greater throughput.
  • Direct scalability from UPLC™, UHPLC, or HPLC screening to purification.
  • Robust chromatographic particles designed for purification.
  • Consistent performance with patented Optimum Bed Density (OBD) design, eliminating voids and extending column life.
  • Scalability assured by bed densities matching analytical columns, reducing method redevelopment.
Catalog No. 50-465-993