Learn More
Waters Corp SunFire C8 OBD Prep Column, 100Å, 10 µm, 50 mm X 250 mm, 1/pk
Supplier:  Waters Corp 186002768
Encompass_Preferred
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
Catalog No. 50-781-359
Only null left
Add to Cart
Request a Quote