Learn More
Waters Corp Waters Preparative Column, C8, OBD, 5 μm, 30 x 150 mm
Supplier:  Waters Corp 186002795
Encompass_Preferred
The SunFire C8 OBD Preparative Column offers efficient, lab-scale separations with excellent column lifetime. Its Optimum Bed Density (OBD) technology ensures uniform packing, enhancing productivity and recovery. The C8 chemistry suits strongly hydrophobic compounds.
  • Uniform, optimum packed bed density.
  • Direct scalability from analytical to preparative LC.
  • Excellent column lifetime.
  • Consistent performance, good resolution, peak shape, and mass loading.
  • Patented OBD design prevents voids and resists bed failure.
  • Designed for high mass loading applications.
Catalog No. 50-781-366
Add to Cart
Request a Quote