Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Prep Column, Silica, OBD, 100Å, 5 μm Particle

Supplier:  Waters Corp 186003436

Encompass_Preferred

The SunFire Prep OBD Silica columns are preparative columns specifically designed for use under normal phase conditions. They feature an Optimum Bed Density (OBD) hardware format, which significantly extends column lifetime and ensures robust performance across a wide range of operating parameters.

  • Optimum Bed Density (OBD) hardware provides superior column lifetime and robust performance.
  • Normal-phase separation mode for chromatographic applications.
  • High silanol activity for specific chromatographic selectivity.
  • Broad pH range of 2 to 8 for versatile sample matrices.
  • High maximum pressure (6000 psi / 415 Bar) compatible with preparative HPLC systems.
  • Spherical silica particle shape (5 μm) ensures efficient separations.
  • 100 Å pore size optimized for a range of analytes.

Catalog No. 50-781-769


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.