Learn More
Waters Corp Preparative Column, C18, 10 μm, 30 × 150 mm
Supplier:  Waters Corp 186003448
Encompass_Preferred
This preparative column is designed to enhance performance in Prep LC by offering uniform, optimum packed bed density. It provides direct scalability from analytical to preparative chromatography, ensuring excellent column lifetime and consistent performance for good resolution, peak shape, and mass loading.
  • Offers uniform, optimum packed bed density.
  • Provides direct scalability from analytical to preparative chromatography.
  • Ensures excellent column lifetime.
  • Delivers consistent column-to-column performance.
  • Leads to good resolution, peak shape, and mass loading.
  • Utilizes traditional stainless steel OBD column technology.
  • Features C18 reversed-phase chemistry.
Catalog No. 50-781-781
Add to Cart
Request a Quote