Learn More
Selleck Chemical LLC Sunitinib 50mg 557795-19-4 SU11248
SDP
Supplier:  Selleck Chemical LLC S778150MG
Encompass_Preferred
Sunitinib (SU11248) is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFR? with IC50 of 80 nM and 2 nM, and also inhibits c-Kit. Sunitinib is also a dose-dependent inhibitor of the autophosphorylation activity of IRE1?. Sunitinib induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1203725