Learn More
Medchemexpress LLC Sunitinib | 557795-19-4 | MFCD09260778 | 99.04% | 398.5 g/mol | 200 MG
Supplier: Medchemexpress LLC 200MGHY10255A
Sunitinib is a multi-targeted receptor tyrosine kinase inhibitor and a small-molecule member of pyrroles and monocarboxylic acid amides. It acts as an antineoplastic agent, an angiogenesis inhibitor, and an immunomodulator. This ATP-competitive inhibitor effectively blocks autophosphorylation of Ire1α, preventing consequent RNase activation.
- Targets VEGFR2 with an IC50 of 80 nM
- Targets PDGFRβ with an IC50 of 2 nM
- Inhibits EC 2.7.10.1 (receptor protein-tyrosine kinase)
- Functions as a neuroprotective agent
- Acts as a vascular endothelial growth factor receptor antagonist
- Approved for treating renal cell carcinoma (RCC) and imatinib-resistant gastrointestinal stromal tumor (GIST)
- Approved for adjuvant treatment of recurrent RCC and pancreatic neuroendocrine tumors (pNET)