Learn More
Selleck Chemical LLC Sunitinib malate S1042-1g
SDP
Supplier:  Selleck Chemical LLC S10421G
Encompass_Preferred
Sunitinib malate is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFR with IC50 of 80 nM and 2 nM in cell-free assays and also inhibits c-Kit Sunitinib Malate effectively inhibits autophosphorylation of Ire1 Sunitinib Malate increases both death receptor and mitochondrial-dependent apoptosis
Catalog No. 50-311-1249