Learn More
Medchemexpress LLC Sunvozertinib | 2370013-49-1 | 99.1% | C29H35ClFN7O3 | 10MG

Supplier: Medchemexpress LLC HY132842A10MG
(S)-Sunvozertinib is the S-enantiomer of sunvozertinib, an irreversible inhibitor of EGFR and BTK used as a research compound to investigate EGFR exon 20 insertion mutations and related oncogenic signaling. It is supplied as a high-purity solid with defined storage conditions for stability.
- High purity suitable for in vitro and in vivo studies.
- Confirmed molecular formula and molecular weight for analytical characterization.
- Multiple package sizes available to support pilot studies and scale-up experiments.
- SMILES provided to support computational chemistry and cheminformatics workflows.
- Storage conditions specified to preserve compound integrity over time.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.