Learn More
MSC Metro Super Adjustable Super Erecta Wire Shelf, Steel, Chrome Plated, 24 in. x 42 in., 1 ea.
Supplier:  MSC 03030624
Encompass
The Super Adjustable Super Erecta chrome wire shelf is an adjustable storage component designed for dry storage environments. Constructed from steel with a chrome-plated finish, this shelf integrates with the existing system of shelves and accessories to provide a rigid and versatile organizational solution.
  • Features a tool-free corner release system for quick, 1 in. (25 mm) incremental adjustments along the post height.
  • Supports a weight capacity of 800 lb. (363 kg) when evenly distributed.
  • Designed to maximize vertical space utilization, potentially increasing storage capacity by 25% or more.
  • Compatible with the existing system of wire shelves and accessories.
  • Includes four pairs of split sleeves for mounting.
  • NSF certified for quality and sanitation standards.
Catalog No. 50-005-11440