Learn More
Bioworld Super Amplifying IF Kit w/ Amplifier Eclipse, 1 Kit
Supplier:  Bioworld 217600101
Encompass_Preferred
This Super Amplifying IF Kit offers the Antibody Amplifier Eclipse™, 1 500ml bottle of Amplifying Antibody DilutionBuffer ™, 1 500ml bottle of 10X Amplifying IHC Wash Buffer™ and 1 250ml bottle of 10X Epitope Unmasking Solution.
Catalog No. 50-199-082