Learn More
Bioworld Super Amplifying IHC Kit w/ Amplifier, 1 Kit
Supplier:  Bioworld 217600091
Encompass_Preferred
This Super Amplifying IHC Kit offers the classic Antibody Amplifier™, 1 500ml bottle of Amplifying Antibody Dilution Buffer ™, 1 500ml bottle of 10X Amplifying IHC Wash Buffer™ and 1 250ml bottle of 10X Epitope Unmasking Solution.
Catalog No. 50-199-081