Learn More
Alkali Scientific Super Broth, Liquid, 1 Liter
SDP
Supplier:  Alkali Scientific SUP002CP
Encompass_Preferred
Super Broth, Liquid, 1 Liter, is an enriched medium designed to provide optimal growth conditions for a wide variety of microorganisms. It contains additional nutrients and vitamins that support rapid cell growth, making it ideal for high-density microbial cultures. This broth is commonly used in microbiological research and biotechnology applications where enhanced cell growth is necessary, such as protein expression, fermentation, and metabolic studies. Super Broth supports a broad range of microorganisms and is widely used in molecular biology labs for cloning, plasmid propagation, and recombinant protein production, offering superior growth conditions for many bacterial strains.
Catalog No. 50-006-60481