Learn More
Leica Biosystems Super Mega Cassette Clamp - Large cassette clamp with adapter for HistoCore microtomes
Supplier:  Leica Biosystems 14050238967
Encompass
The Super Mega cassette clamp is a larger clamp assembly designed to secure oversized tissue cassettes during microtomy. It provides a broader clamping surface and includes an adapter to accommodate large-format cassettes; listed as an optional accessory for HistoCore series microtomes.
  • Holds large-format tissue cassettes.
  • Includes adapter for large cassettes.
  • Dimensions (HxWxD): 75 x 52 x 35 mm.
  • Silver finish.
  • Supplied as an optional accessory for HistoCore microtomes.
Catalog No. NC2820694