Learn More
Cole-Parmer SUPER FLANGELESS FERRULE GREEN
Supplier:  Cole-Parmer 0202141
Encompass
Super Flangeless Ferrule, Green ETFE/316 SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK
Catalog No. NC2186221