Learn More
Uline SUPER GRIPTION HI SZ L 12PR/CS
Supplier:  Uline S19885L
Encompass
APPEARANCE:Overedge Color: BrownTHICKNESS:10 gaugeMATERIAL:Coating: Sandy LatexSilicone freeCLEANING INSTRUCTIONS: Use tempered water not to exceed 104° F for 10 minutes. Rinse with tempered water & tumble dry at a maximum 104° F. SPECIFICATIONS:Standard ANSI/ISEA 105-2016:Cut Level: A2Abrasion Level: 3
Catalog No. NC2819297