Learn More
Cusabio Technology LLC Mouse Super Oxidase DimutaseSOD ELISA Kit
Supplier:  Cusabio Technology LLC CSBE08556M
Encompass_Preferred
The Mouse Super Oxidase Dimutase (SOD) ELISA Kit allows for the detection and quantification of SOD in mouse serum, plasma, or tissue homogenates. This kit has high sensitivity, excellent specificity, precision less than 10%, and consistency between batches. It employs the sandwich ELISA mechanism and enzyme-substrate chromogenic reaction to achieve the detection. The solution color develops in proportion to the amount of SOD in the sample, and the color intensity can be measured at 450 nm via a microplate reader.
Catalog No. NC3654208