Learn More
Biogenex Laboratories Super SensitiveTM (SS) Link-Label IHC Detection System
SDP
Supplier:  Biogenex Laboratories QP9009LE
Encompass_Preferred
A classic system based on the highly specific and sensitive streptavidin-biotin interaction to detect a bound antibody. These kits include multi-Link – a mix of anti-mouse and anti-rabbit IgGs conjugated to multiple biotin molecules and a Label -Strepavidin conjugated with an enzyme (horseradish peroxidase (HRP) or alkaline phosphatase (AP))
Catalog No. 50-223-8110