Learn More
Immuno4 LLC SUPER SHIELD HRP STABILIZER 1L
Supplier:  Immuno4 LLC HRPZR
Encompass
Bovine and Mammalian Protein Free Stabilizer, One Component Ready to Use. Eliminates BSE regulatory concerns. This superior stabilizer was developed to maximize the life of your conjugates while eliminating troublesome interactions.
Catalog No. NC0680010