Learn More
Medchemexpress LLC HY-P1728A 1mg Medchemexpress, Super-TDU (1-31) (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1728A1MG
Encompass_Preferred
Medchemexpress, HY-P1728A 1mg Super-TDU (1-31) (TFA) CAS: Super-TDU (1-31) is a peptide of Super-TDU, which is an inhibitor of YAP-TEADs , shows potent anti-tumor activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4242