Learn More
Sigma Aldrich Fine Chemicals Biosciences SuperdexR 75 Prep Grade Cy
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17104401
Encompass
Superdex(R) 75 prep grade gel filtration medium offers very high reSOLUTIONon fractionation for separation polishing and product formulation of recombinant proteins.Superdex(R) prep grade is a preparative gel filtration medium with a composite matrix of dextran and agarose. This matrix combines the excellent gel filtration properties of cross-linked dextran (Sephadex) with the physical and chemical stabilities of highly cross-linked agarose to produce a separation medium with outstanding selectivity and high reSOLUTIONon. In addition its low non-specific interaction permits high recovery of biological material. Together these properties make Superdex(R) prep grade the first choice gel filtration media for all applications from laboratory to process scale.Superdex(R) prep grade is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.As a BioProcess medium
Catalog No. 50-290-8071