Learn More
Sino Biological SuperNuclease (50% Glycerol) 500 kU
Supplier:  Sino Biological GMPSSNP01G500
Encompass_Preferred
SuperNuclease is a recombined Serratia marcescens’ extracellular endonuclease. The sequence is same with Benzonase (Benzonase is a trademark of Merck KGaA, Darmstadt, Germany).
Catalog No. 50-006-96232