Learn More
Cusabio Technology LLC Fish Superoxide Dismutase(SOD)ELISA kit
Supplier:  Cusabio Technology LLC CSBE15929FH
Encompass_Preferred
This Fish SOD ELISA Kit was designed for the quantitative measurement of Fish SOD protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 1.25 ng/mL-20 ng/mL and the sensitivity is 0.625 ng/mL. One kit contains 96 wells.
Catalog No. NC2952965