Learn More
LIFE TECH (BID ECHO ONLY) SuperScript™ IV RT Reaction Buffer
Supplier:  LIFE TECH (BID ECHO ONLY) 18090050B
Encompass
SuperScript IV RT Reaction Buffer is included with, and designed for use with, SuperScript IV Reverse Transcriptase. This buffer is optimized to give robust and reliable cDNA synthesis. In cases where additional quantity is required, SuperScript IV RT Reaction Buffer is made available here in a stand-alone format.
Catalog No. NC3859469