Learn More
FUJIFILM BIOSCIENCES INC SuperSep(TM) Ace Mini, 10-20%, 17well, 10ea per pack
Supplier:  FUJIFILM BIOSCIENCES INC 19118613
Encompass_Preferred
SuperSep Ace Mini is polyacrylamide gel for protein electrophoresis. This is even smaller than commom minigel. Using this can finish electrophoresis in 20 min.
Catalog No. NC3740632