Learn More
Medchemexpress LLC HY-17002 100mg Medchemexpress, Suplatast (Tosilate) CAS:94055-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17002100MG
Encompass_Preferred
Medchemexpress, HY-17002 100mg Suplatast (Tosilate) CAS:94055-76-2 Suplatast Tosilate (IPD 1151T) is an orally active Th2 cytokine inhibitor which can inhibit both IL-4 and IL-5 production from Th2 cells and suppress IgE synthesis. Suplatast Tosilate is an anti-allergic agent. Suplatast Tosilate has antiasthmatic, anti-inflammatory and antifibrotic activity[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1924