Learn More
Selleck Chemical LLC Suplatast Tosylate S2015-25mg
SDP
Supplier:  Selleck Chemical LLC S201525MG
Encompass_Preferred
Suplatast Tosylate (IPD-1151T Suplatast Tosilate) is a novel capsular anti-asthmatic agent that suppresses both IgE production IL-4 and IL-5 synthesis with IC50 above 100 M
Catalog No. 50-311-0382