Learn More
Health Care Logistics Inc Supor Sterile Syringe Filter, 5 Micron, 32Mm
SDP
Supplier:  Health Care Logistics Inc 12554
Encompass
Designed to assure quality control of sterile drugs. Hydrophilic Supor membrane provides superior throughputs and flow rates over conventional filter devices. Double luer lock design for secure needle and syringe attachment.
Catalog No. NC1431248