Learn More
APICAL CONSULTATIONS SUPPLEMENTARY 5L/10L TW JLR
Supplier:  APICAL CONSULTATIONS CUSTOMSYSTEM/Q1309
Encompass
Supplementary 5 & 10L TW JLR Suite Components:Wetted Materials: Borosilicate 3.3, virgin PTFE/ePTFE, Hastelloy C22 & FFKMSS316 castered structure & hardware (interchangeable lift/lower system: 5 & 10L vessels) -50 to 150C0-14 pHFull Vacuum (<50torr) to 1.0 barDesign Pressure: 1.25 bar-2x triple-wall DN300 5L vessel-2x triple-wall DN300 10L vessel-2x Hastelloy C22 lid-3x FFKM o-ring (lid)-3x PTFE PT100 baffle-type sensor w/ Hastelloy tip for in-liquid temp-2x ATEX PT100 for vapor temp-2x jacket transfer fluid adapter sets w/ dry break-2x SS316 castered structure w/ shielding-2x Hastelloy Premex Miniluxe mechanical seal w/ integrated gas management-2x SS316 receiver cooling bath-2x C22 Hastelloy retreat curve stir prop-2x C22 Hastelloy anchor stir prop-2x SS316 control panel-2x set of nozzles per approved drawing-2x vacuum liquid sampling system
Catalog No. NC3938159