Learn More
BRANSON ULTRASONICS CORPORATION (3P) SUPPORT RACK FOR FS110
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPN916042
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Support Racks, For Model 5800, For Use With: For Model 5800, Support rack, Height: 15 in., 38.1cm, Width: 15 in.
Catalog No. NC3721423