Learn More
Sigma Aldrich Fine Chemicals Biosciences SuRE/Cut Buffer H
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11417991001/DEL/DEL
Encompass
The SuRE/Cut Buffer System consists of five optimized incubation buffers, 10x concentrated, for DNA restriction digest. Activity of all restriction enzymes has been determined in each buffer to select 100% activity or to calculate activity in double-digests. The SuRE/Cut Buffer H has been used for digestion of genomic DNA using the restriction enzyme XbaI. It has been used for digestion of PCR products and plasmid using EcoRI and XhoI.
Catalog No. NC0979923