Learn More
Cayman Chemical SureLIghtB-PhycoerythrInB 10mg
SDP
Supplier:  Cayman Chemical 1663210 MG
Encompass_Preferred
Because of its high quantum yield, B-PE is considered the world’s brightest fluorophore. It is compatible with commonly available lasers and gives exceptional results in flow cytometry, Luminex and immunofluorescent staining. B-PE is also less “sticky” than common synthetic fluorophores and therefore gives less background interference. Uses: Flow cytometry, Fluorescence-activated cell sorting, Custom Antibody labeling, Luminex, and T Cell labeling
Catalog No. 50-296-6735