Learn More
AUTOMATIONDIRECT COM INC SURESTEP EXTENSION CABLE
Supplier:  AUTOMATIONDIRECT COM INC STPEXT010
Encompass
SureStep extension cable, 4-pin connector to pigtail, 10ft cablelength. For use with SureStep MTR series stepper motors.
Catalog No. NC3066473