Learn More
Electron Microscopy Sciences EMS Surface Mount Tweezers Style SM 108, 120 mm
SDP
Supplier:  Electron Microscopy Sciences 7825010
Encompass_Preferred
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Tips are cut on a 45 degree angle with a 1.5mm inside gripping groove. Ideal for positioning.
Catalog No. 50-239-19