Learn More
Medchemexpress LLC Surfactin | 252023-70-4 | 99.0% | 1025.04 g/mol | No single molecular formula (mixture of isomeric surfactins) | 5 MG

Supplier: Medchemexpress LLC HY1295555MG
Surfactin is a cyclic lipopeptide biosurfactant (mixture of isomeric forms) used in research on antimicrobial, antifungal, and antiviral activity. Supplied as a high-purity research reagent, it includes documented solubility and storage recommendations for in vitro and in vivo studies.
- High purity: 98.96%.
- Average molecular weight: 1025.04 g/mol.
- Soluble in water and DMSO with ultrasonic assistance.
- Powder storage: stable at -20°C for long term.
- Commonly used in antimicrobial, antiviral, and surfactant research.
- Available in small research pack sizes from 5 mg to larger quoted quantities.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.