Learn More
Med Vet International Surgical Instrument Guards , Black, 1/8" x 1" x 1", J0926J, 50 Count
Supplier:  Med Vet International JOR926J
Encompass_Preferred
JorVet instrument guards are designed to protect valuable instrument tips from damage during sterilization and handling. Sharp and pointed tips can break through sterilization pouches and injure the surgeon. 50/pkg.Non-sterile, but can be autoclaved or gassterilizedMedical-grade plastic (vented)Guards can also work as covers over exposed bone pins.
Catalog No. 50-005-83999