Learn More
Medchemexpress LLC Surufatinib (sulfatinib) | 1308672-74-3 | >98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY122971MG
Encompass_Preferred
Sulfatinib (surufatinib) is a small-molecule tyrosine kinase inhibitor that selectively targets VEGFR1/2/3, FGFR1, and CSF1R. It is supplied as a high-purity analytical reference for research into angiogenesis and tumor signaling pathways.
  • Targets VEGFR1/2/3, FGFR1, and CSF1R with low-nanomolar potency.
  • High purity analytical standard (≥98%).
  • Molecular formula C24H28N6O3S and molecular weight 480.6 g·mol-1.
  • Solid powder formulation suitable for analytical and in vitro research.
  • Available in small quantities for laboratory use (e.g., 1 MG).
Catalog No. 50-002-30524